accessibility menu, dialog, popup

UserName

CAS RN 3160-35-8

IUPAC Name: (3E)-4-(4-hydroxyphenyl)but-3-en-2-one
Synonyms: 4-hydroxycinnamoylmethane
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: OCNIKEFATSKIBE-NSCUHMNNSA-N
SMILES: CC(=O)\C=C\C1=CC=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results