accessibility menu, dialog, popup

UserName

CH3IH3C

CAS RN 31599-61-8

IUPAC Name: 4-iodo-1,2-dimethylbenzene
Synonyms: 4-iodo-1,2-dimethylbenzene
Molecular Weight (g/mol): 232.06
Molecular Formula: C8H9I
InChi Key: CSFRCLYFVINMBZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(I)C=C1C
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)