accessibility menu, dialog, popup

UserName

CAS RN 31566-31-1

H3COOOHOH

CAS RN 31566-31-1

IUPAC Name: 2,3-dihydroxypropyl octadecanoate
Synonyms: glyceryl stearate grocor 5500 emcol msk lipo gms 600 dermagine lipo gms 450 lipo gms 410 drewmulse TP monostearate (glyceride) abracol S.L.G.
Molecular Weight (g/mol): 358.56
Molecular Formula: C21H42O4
InChi Key: VBICKXHEKHSIBG-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results