accessibility menu, dialog, popup

UserName

CAS RN 3156-34-1

ONOCl(E)

CAS RN 3156-34-1

IUPAC Name: 1-chloro-2-[(1E)-2-nitroethenyl]benzene
Synonyms: 2-chloro-B-nitrostyrene
Molecular Weight (g/mol): 183.59
Molecular Formula: C8H6ClNO2
InChi Key: QHKJTRDWAZGBLR-AATRIKPKSA-N
SMILES: [O-][N+](=O)\C=C\C1=CC=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results