accessibility menu, dialog, popup

UserName

ONOONOO

CAS RN 31551-45-8

IUPAC Name: 2,7-dinitro-9H-fluoren-9-one
Synonyms: 9H-fluoren-9-one, 2,7-dinitro-
Molecular Weight (g/mol): 270.2
Molecular Formula: C13H6N2O5
InChi Key: HDVGAFBXTXDYIB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2C3=CC=C(C=C3C(=O)C2=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results