accessibility menu, dialog, popup

UserName

CAS RN 3147-62-4

IUPAC Name: 3,5-dihydroxybenzamide
Synonyms: 3,5-dihydroxybenzamide
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: PLYYMFBDRBSPJZ-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(O)=CC(O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results