accessibility menu, dialog, popup

UserName

CH3H3CNONOO

CAS RN 313496-12-7

IUPAC Name: N-benzyl-4-nitro-N-(propan-2-yl)benzamide
Synonyms: N-benzyl-N-isopropyl-4-nitrobenzamide
Molecular Weight (g/mol): 298.34
Molecular Formula: C17H18N2O3
InChi Key: WOIOOHBTONFDSI-UHFFFAOYSA-N
SMILES: CC(C)N(CC1=CC=CC=C1)C(=O)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results