accessibility menu, dialog, popup

UserName

NaNH2IIOOI

CAS RN 3119-15-1

IUPAC Name: sodium 3-amino-2,4,6-triiodobenzoate
Synonyms: sodium 3-amino-2,4,6-triiodobenzoate
Molecular Weight (g/mol): 536.81
Molecular Formula: C7H3I3NNaO2
InChi Key: CTXRMWPLTDAHOR-UHFFFAOYSA-M
SMILES: [Na+].NC1=C(I)C=C(I)C(C([O-])=O)=C1I
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)