accessibility menu, dialog, popup

UserName

CAS RN 3109-63-5

H3CNCH3H3CH3CFPFFFFF

CAS RN 3109-63-5

IUPAC Name: hexafluoro-λ⁵-phosphanuide; tetrabutylazanium
Synonyms: tetrabutylammonium hexafluorophosphate
Molecular Weight (g/mol): 387.44
Molecular Formula: C16H36F6NP
InChi Key: BKBKEFQIOUYLBC-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)