accessibility menu, dialog, popup

UserName

NNClClH2N

CAS RN 310400-38-5

IUPAC Name: 5,6-dichloropyrimidin-4-amine
Synonyms: 5,6-dichloropyrimidin-4-amine
Molecular Weight (g/mol): 163.99
Molecular Formula: C4H3Cl2N3
InChi Key: ISTJUVRUUBNZNB-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C(Cl)=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results