accessibility menu, dialog, popup

UserName

CAS RN 31008-64-7

N(R)N(R)O(S)(R)NNOHNH2HOOPOPOOOOOLiLiLi

CAS RN 31008-64-7

IUPAC Name: trilithium(1+) ({[5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Synonyms: trilithium(1+) {[5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxyphosphonate
Molecular Weight (g/mol): 445
Molecular Formula: C10H12Li3N5O10P2
InChi Key: LZGPPAHUZSOGHJ-UHFFFAOYNA-K
SMILES: [Li+].[Li+].[Li+].NC1=C2N=CN(C3OC(COP([O-])(=O)OP([O-])([O-])=O)C(O)C3O)C2=NC=N1
Physical Form 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)