accessibility menu, dialog, popup

UserName

H3COOH3CH2SiSH

CAS RN 31001-77-1

IUPAC Name: 3-[(dimethoxymethyl)silyl]propane-1-thiol
Synonyms: 3-[(dimethoxymethyl)silyl]propane-1-thiol
Molecular Weight (g/mol): 180.34
Molecular Formula: C6H16O2SSi
InChi Key: LOOUJXUUGIUEBC-UHFFFAOYSA-N
SMILES: COC(OC)[SiH2]CCCS
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results