CAS RN 30992-81-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3’-Hydroxy Warfarin, TRC
CAS: 30992-81-5 Molecular Formula: C19 H16 O5 Molecular Weight (g/mol): 324.33 Synonym: 4-Hydroxy-3-[1-(3-hydroxyphenyl)-3-oxobutyl]-2H-1-benzopyran-2-one,3-(α-Acetonyl-m-hydroxybenzyl)-4-hydroxycoumarin IUPAC Name: 4-hydroxy-3-[1-(3-hydroxyphenyl)-3-oxo-butyl]chromen-2-one SMILES: CC(=O)CC(C1=C(O)c2ccccc2OC1=O)c3cccc(O)c3