accessibility menu, dialog, popup

UserName

CAS RN 3096-81-9

NO

CAS RN 3096-81-9

IUPAC Name: 4-phenoxybenzonitrile
Synonyms: 4-phenoxybenzonitrile
Molecular Weight (g/mol): 195.22
Molecular Formula: C13H9NO
InChi Key: UYHCIOZMFCLUDP-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(OC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)