accessibility menu, dialog, popup

UserName

H3CNCH3NCH3NCH3NCH3CH3

CAS RN 3083-10-1

IUPAC Name: 2,5,8,11-tetramethyl-2,5,8,11-tetraazadodecane
Synonyms: 2,5,8,11-tetramethyl-2,5,8,11-tetraazadodecane
Molecular Weight (g/mol): 230.4
Molecular Formula: C12H30N4
InChi Key: DWFKOMDBEKIATP-UHFFFAOYSA-N
SMILES: CN(C)CCN(C)CCN(C)CCN(C)C
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)