accessibility menu, dialog, popup

UserName

CAS RN 30766-11-1

OOBrN

CAS RN 30766-11-1

IUPAC Name: 5-bromopyridine-2-carboxylate
Synonyms: 5-bromopyridine-2-carboxylate
Molecular Weight (g/mol): 201
Molecular Formula: C6H3BrNO2
InChi Key: MNNQIBXLAHVDDL-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(Br)C=N1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)