CAS RN 307308-03-8
CAS RN 307308-03-8
IUPAC Name:
4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-3-carbaldehyde
Synonyms:
4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-3-carbaldehyde
Molecular Weight (g/mol):
150.18
Molecular Formula:
C8H10N2O
InChi Key:
CSPINDKIWMCRNX-UHFFFAOYSA-N
SMILES:
O=CC1=C2CCCCN2N=C1
1
–
1
of
1
results
1
4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde, 97%, Thermo Scientific Chemicals
CAS: 307308-03-8 Molecular Formula: C8H10N2O Molecular Weight (g/mol): 150.18 InChI Key: CSPINDKIWMCRNX-UHFFFAOYSA-N Synonym: 4,5,6,7-tetrahydropyrazolo 1,5-a pyridine-3-carbaldehyde,4,5,6,7-tetrahydropyrazolo 1,5-a pyridine-3-carboxaldehyde,4h,5h,6h,7h-pyrazolo 1,5-a pyridine-3-carbaldehyde,4,5,6,7-tetrahydro-pyrazolo 1,5-a pyridine-3-carbaldehyde,pyrazolo 1,5-a pyridine-3-carboxaldehyde,4,5,6,7-tetrahydro-9ci,pyrazolo 1,5-a pyridine-3-carboxaldehyde, 4,5,6,7-tetrahydro-9ci PubChem CID: 45083680 IUPAC Name: 4,5,6,7-tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde SMILES: C1CCN2C(=C(C=N2)C=O)C1