accessibility menu, dialog, popup

UserName

CAS RN 306937-38-2

CH3NSH3COHO

CAS RN 306937-38-2

IUPAC Name: 2-(2,5-dimethyl-1,3-thiazol-4-yl)acetic acid
Synonyms: (2,5-dimethyl-1,3-thiazol-4-yl)acetic acid
Molecular Weight (g/mol): 171.21
Molecular Formula: C7H9NO2S
InChi Key: FFPWICPYXBDRHM-UHFFFAOYSA-N
SMILES: CC1=NC(CC(O)=O)=C(C)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results