accessibility menu, dialog, popup

UserName

CAS RN 306937-27-9

CH3H3CCH3NHNH2HCl

CAS RN 306937-27-9

IUPAC Name: (3-tert-butylphenyl)hydrazine hydrochloride
Synonyms: (3-tert-butylphenyl)hydrazine hydrochloride
Molecular Weight (g/mol): 200.71
Molecular Formula: C10H17ClN2
InChi Key: XKLKKAKHNGXDPA-UHFFFAOYSA-N
SMILES: Cl.CC(C)(C)C1=CC(NN)=CC=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results