accessibility menu, dialog, popup

UserName

CAS RN 306937-23-5

IUPAC Name: 1-tert-butyl-5-methyl-1H-pyrazole-3-carbohydrazide
Synonyms: 1-tert-butyl-5-methylpyrazole-3-carbohydrazide
Molecular Weight (g/mol): 196.25
Molecular Formula: C9H16N4O
InChi Key: TWKKUKBETOOWRR-UHFFFAOYSA-N
SMILES: CC1=CC(=NN1C(C)(C)C)C(=O)NN
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results