accessibility menu, dialog, popup

UserName

CAS RN 306937-22-4

H3COONNH2

CAS RN 306937-22-4

IUPAC Name: ethyl 1-[(3-aminophenyl)methyl]piperidine-4-carboxylate
Synonyms: ethyl 1-[(3-aminophenyl)methyl]piperidine-4-carboxylate
Molecular Weight (g/mol): 262.35
Molecular Formula: C15H22N2O2
InChi Key: WXYBQSXOZUFNJY-UHFFFAOYSA-N
SMILES: CCOC(=O)C1CCN(CC2=CC(N)=CC=C2)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results