accessibility menu, dialog, popup

UserName

FBr

CAS RN 306935-88-6

IUPAC Name: 3-bromo-4-fluoro-1,1'-biphenyl
Synonyms: 3-bromo-4-fluoro-1,1'-biphenyl
Molecular Weight (g/mol): 251.1
Molecular Formula: C12H8BrF
InChi Key: COWXPZSVUXHAFS-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results