accessibility menu, dialog, popup

UserName

CAS RN 306935-54-6

OClOCl

CAS RN 306935-54-6

IUPAC Name: 6-chloro-2H-chromene-3-carbonyl chloride
Synonyms: 6-chloro-2H-chromene-3-carbonyl chloride
Molecular Weight (g/mol): 229.06
Molecular Formula: C10H6Cl2O2
InChi Key: HGFGWTNRUGRBNQ-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC2=C(OC1)C=CC(Cl)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results