accessibility menu, dialog, popup

UserName

CAS RN 306934-70-3

NHNNFFF

CAS RN 306934-70-3

IUPAC Name: 1-[5-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane
Synonyms: 1-[5-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane
Molecular Weight (g/mol): 245.25
Molecular Formula: C11H14F3N3
InChi Key: IBMSHQVIAUTKDL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(C=C1)N1CCCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results