CAS RN 3069-25-8
CAS RN 3069-25-8
IUPAC Name:
methyl[3-(trimethoxysilyl)propyl]amine
Synonyms:
methyl[3-(trimethoxysilyl)propyl]amine
Molecular Weight (g/mol):
193.32
Molecular Formula:
C7H19NO3Si
InChi Key:
DVYVMJLSUSGYMH-UHFFFAOYSA-N
SMILES:
CNCCC[Si](OC)(OC)OC
1
–
1
of
1
results
1
(3-Methylaminopropyl)trimethoxysilane, 95%
CAS: 3069-25-8 Molecular Formula: C7H19NO3Si Molecular Weight (g/mol): 193.32 MDL Number: MFCD00048021 InChI Key: DVYVMJLSUSGYMH-UHFFFAOYSA-N Synonym: n-methylaminopropyltrimethoxysilane,n-methyl-3-trimethoxysilyl propan-1-amine,trimethoxy 3-methylamino propyl silane,1-propanamine, n-methyl-3-trimethoxysilyl,n-methyl-3-trimethoxysilyl propylamine,n-methyl-3-aminopropyltrimethoxysilane,3-methylaminopropyl-trimethoxysilane,methyl 3-trimethoxysilyl propyl amine,n-methyl-3-aminopropyl-trimethoxysilane,acmc-20ak27 PubChem CID: 76480 IUPAC Name: N-methyl-3-trimethoxysilylpropan-1-amine SMILES: CNCCC[Si](OC)(OC)OC