accessibility menu, dialog, popup

UserName

CAS RN 3054-92-0

CH3H3CCH3CH3CH3HO

CAS RN 3054-92-0

IUPAC Name: 2,3,4-trimethylpentan-2-ol
Synonyms: 2,3,4-trimethylpentan-2-ol
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: FTPXXARYDFWPGE-UHFFFAOYNA-N
SMILES: CC(C)C(C)C(C)(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results