accessibility menu, dialog, popup

UserName

CH3OOClN

CAS RN 30529-70-5

IUPAC Name: 2-chloro-6-methylpyridine-3-carboxylate
Synonyms: 2-chloro-6-methylpyridine-3-carboxylate
Molecular Weight (g/mol): 170.57
Molecular Formula: C7H5ClNO2
InChi Key: ACQXHCHKMFYDPM-UHFFFAOYSA-M
SMILES: CC1=CC=C(C([O-])=O)C(Cl)=N1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)