accessibility menu, dialog, popup

UserName

H3COOBr

CAS RN 3042-81-7

IUPAC Name: methyl 2-bromo-2-phenylacetate
Synonyms: methyl 2-bromo-2-phenylacetate
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: NHFBYYMNJUMVOT-UHFFFAOYNA-N
SMILES: COC(=O)C(Br)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)