accessibility menu, dialog, popup

UserName

OHN

CAS RN 3040-44-6

IUPAC Name: 2-(piperidin-1-yl)ethan-1-ol
Synonyms: 1-piperidineethanol
Molecular Weight (g/mol): 129.2
Molecular Formula: C7H15NO
InChi Key: KZTWONRVIPPDKH-UHFFFAOYSA-N
SMILES: OCCN1CCCCC1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (3)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)