CAS RN 303727-31-3
IUPAC Name:
2-chloro-5-[2-phenyl-5-(pyridin-4-yl)-1H-imidazol-4-yl]phenol
Synonyms:
2-chloro-5-[2-phenyl-5-(pyridin-4-yl)-1H-imidazol-4-yl]phenol
Molecular Weight (g/mol):
347.8
Molecular Formula:
C20H14ClN3O
InChi Key:
WXJLXRNWMLWVFB-UHFFFAOYSA-N
SMILES:
OC1=C(Cl)C=CC(=C1)C1=C(NC(=N1)C1=CC=CC=C1)C1=CC=NC=C1
1
–
1
of
1
results
1
L-779450, MedChemExpress
MedChemExpress L-779450 is a potent and selective B-Raf kinase inhibitor with a Kd of 2.4 nM.