accessibility menu, dialog, popup

UserName

ClClI

CAS RN 3032-81-3

IUPAC Name: 1,3-dichloro-5-iodobenzene
Synonyms: 1,3-dichloro-5-iodobenzene
Molecular Weight (g/mol): 272.89
Molecular Formula: C6H3Cl2I
InChi Key: AATPRMRVLQZEHB-UHFFFAOYSA-N
SMILES: ClC1=CC(I)=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)