CAS RN 30314-64-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(p-Methoxyphenyl)-2-propanol, TRC
CAS: 30314-64-8 Molecular Formula: C10H14O2 Molecular Weight (g/mol): 166.22 Synonym: 1-(4-Methoxyphenyl)-2-propanol,2-Hydroxy-1-(4-methoxyphenyl)propane,2'-Hydroxy-p-propylanisole,p-Methoxy-alpha-methylphenethyl Alcohol,4-Methoxy-alpha-methylbenzeneethanol IUPAC Name: 1-(4-methoxyphenyl)propan-2-ol SMILES: COc1ccc(CC(C)O)cc1