accessibility menu, dialog, popup

UserName

CAS RN 303136-82-5

H3CCH3NN

CAS RN 303136-82-5

IUPAC Name: 2,9-dimethyl-1,10-phenanthroline
Synonyms: neocuproine
Molecular Weight (g/mol): 208.26
Molecular Formula: C14H12N2
InChi Key: IYRGXJIJGHOCFS-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results