accessibility menu, dialog, popup

UserName

CAS RN 302911-97-3

HOFFFFFF

CAS RN 302911-97-3

IUPAC Name: [2,5-bis(trifluoromethyl)phenyl]methanol
Synonyms: [2,5-bis(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 244.14
Molecular Formula: C9H6F6O
InChi Key: IFJMDKFBPFFXPC-UHFFFAOYSA-N
SMILES: OCC1=CC(=CC=C1C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results