accessibility menu, dialog, popup

UserName

CAS RN 30273-42-8

CH3H2NClBr

CAS RN 30273-42-8

IUPAC Name: 4-bromo-2-chloro-6-methylaniline
Synonyms: 4-bromo-2-chloro-6-methylaniline
Molecular Weight (g/mol): 220.49
Molecular Formula: C7H7BrClN
InChi Key: DIXGIKZIIZRFKE-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC(Cl)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results