CAS RN 3025-96-5
CAS RN 3025-96-5
IUPAC Name:
4-acetamidobutanoic acid
Synonyms:
4-acetamidobutyrate 4-(acetylamino)butyrate
Molecular Weight (g/mol):
145.16
Molecular Formula:
C6H11NO3
InChi Key:
UZTFMUBKZQVKLK-UHFFFAOYSA-N
SMILES:
CC(=O)NCCCC(O)=O
1
–
2
of
2
results
1
4-Acetamidobutanoic acid, MedChemExpress
MedChemExpress 4-Acetamidobutanoic acid (N-acetyl GABA), the main metabolite of GABA, exhibits antioxidant and antibacterial activities.
2
4-Acetamidobutyric acid, 99%, Thermo Scientific™
CAS: 3025-96-5 Molecular Formula: C6H11NO3 Molecular Weight (g/mol): 145.158 MDL Number: MFCD00020552 InChI Key: UZTFMUBKZQVKLK-UHFFFAOYSA-N Synonym: 4-acetamidobutyric acid,4-acetylamino butanoic acid,n-acetyl-4-aminobutyric acid,n-acetyl-4-aminobutanoic acid,n-acetyl-gamma-aminobutyrate,butyric acid, 4-acetamido,4-acetamidobutanoate,4-acetamidobutyrate,gamma-acetylaminobutyric acid,n4-acetylaminobutanoate PubChem CID: 18189 ChEBI: CHEBI:17645 IUPAC Name: 4-acetamidobutanoic acid SMILES: CC(=O)NCCCC(=O)O