Learn More
4-Acetamidobutanoic acid, MedChemExpress
Product Code. 16470926
Encompass_Preferred
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
Product Code. 16470926
Brand: Medchem Express HY101411
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress 4-Acetamidobutanoic acid (N-acetyl GABA), the main metabolite of GABA, exhibits antioxidant and antibacterial activities.
4-Acetamidobutanoic acid (N-acetyl GABA), the main metabolite of GABA, exhibits antioxidant and antibacterial activities.
Specifications
Chemical Name or Material 4-Acetamidobutanoic acid
Physical Form Solid
CAS 3025-96-5
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C6H11NO3
Synonym N-acetyl GABA
Solubility Information H2O : 50 mg/mL (344.45 mM; Need ultrasonic)
SMILES O=C(O)CCCNC(C)=O
Molecular Weight (g/mol) 145.16
Formula Weight 145.16
Percent Purity 98.0%
Grade Research