accessibility menu, dialog, popup

UserName

CAS RN 30235-27-9

NH2SNN

CAS RN 30235-27-9

IUPAC Name: 4-(pyridin-3-yl)-1,3-thiazol-2-amine
Synonyms: 4-(pyridin-3-yl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 177.23
Molecular Formula: C8H7N3S
InChi Key: XOHZQGAYUHOJPR-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results