accessibility menu, dialog, popup

UserName

H3COCH3H3CBOHHO

CAS RN 301699-39-8

IUPAC Name: (4-methoxy-3,5-dimethylphenyl)boronic acid
Synonyms: 4-methoxy-3,5-dimethylphenylboronic acid
Molecular Weight (g/mol): 180.01
Molecular Formula: C9H13BO3
InChi Key: WZUCSPWZHRVOSD-UHFFFAOYSA-N
SMILES: COC1=C(C)C=C(C=C1C)B(O)O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results