accessibility menu, dialog, popup

UserName

CH3H3CCH3OONI

CAS RN 301673-14-3

IUPAC Name: tert-butyl 4-iodopiperidine-1-carboxylate
Synonyms: tert-butyl 4-iodopiperidine-1-carboxylate
Molecular Weight (g/mol): 311.16
Molecular Formula: C10H18INO2
InChi Key: YFWQFKUQVJNPKP-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(I)CC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)