accessibility menu, dialog, popup

UserName

CAS RN 3010-57-9

H3CNCH3NNCH3(E/Z)

CAS RN 3010-57-9

IUPAC Name: N,N-dimethyl-4-[2-(4-methylphenyl)diazen-1-yl]aniline
Synonyms: N,N-dimethyl-4-[2-(4-methylphenyl)diazen-1-yl]aniline
Molecular Weight (g/mol): 239.32
Molecular Formula: C15H17N3
InChi Key: IPIZWPBCCYXHDW-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results