CAS RN 300827-87-6
CAS RN 300827-87-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Desvenlafaxine Hydrochloride, TRC
CAS: 300827-87-6 Molecular Formula: C16 H25 N O2 . Cl H Molecular Weight (g/mol): 299.84 Synonym: O-Desmethylvenlafaxine hydrochloride,Phenol, 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]-, hydrochloride (1:1),Phenol, 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]-, hydrochloride (9CI),4-[2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol hydrochloride,Desvenlafaxine hydrochloride,O-Desvenlafaxine hydrochloride IUPAC Name: 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol;hydrochloride SMILES: Cl.CN(C)CC(c1ccc(O)cc1)C2(O)CCCCC2