accessibility menu, dialog, popup

UserName

H3CCH3OCH3O

CAS RN 3002-23-1

IUPAC Name: 6-methylheptane-2,4-dione
Synonyms: 6-methylheptane-2,4-dione
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: IGMOYJSFRIASIE-UHFFFAOYSA-N
SMILES: CC(C)CC(=O)CC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results