accessibility menu, dialog, popup

UserName

CAS RN 29956-99-8

CH3CH3H3CCH3H3CSSCH3H3CCH3CH3H3C

CAS RN 29956-99-8

IUPAC Name: 2,2,4-trimethyl-4-[(2,4,4-trimethylpentan-2-yl)disulfanyl]pentane
Synonyms: 2,2,4-trimethyl-4-[(2,4,4-trimethylpentan-2-yl)disulfanyl]pentane
Molecular Weight (g/mol): 290.57
Molecular Formula: C16H34S2
InChi Key: ZIMCZOLRXKPXLN-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)SSC(C)(C)CC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results