accessibility menu, dialog, popup

UserName

CAS RN 2991-28-8

IUPAC Name: 2,5-difluorobenzoic acid
Synonyms: 2,5-difluorobenzoic acid
Molecular Weight (g/mol): 158.1
Molecular Formula: C7H4F2O2
InChi Key: LBQMIAVIGLLBGW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=CC=C1F
Molecular Weight (g/mol) 
  • (3)
  • (6)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (6)