accessibility menu, dialog, popup

UserName

H3COSOOOCH3NN

CAS RN 299-11-6

IUPAC Name: 5-methylphenazin-5-ium methyl sulfate
Synonyms: phenazine methosulfate methylphenazonium methosulfate phenazine methosulphate
Molecular Weight (g/mol): 306.34
Molecular Formula: C14H14N2O4S
InChi Key: RXGJTUSBYWCRBK-UHFFFAOYSA-M
SMILES: COS([O-])(=O)=O.C[N+]1=C2C=CC=CC2=NC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (1)
Quantity 
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)