accessibility menu, dialog, popup

UserName

CAS RN 2987-53-3

H3CSH2N

CAS RN 2987-53-3

IUPAC Name: 2-(methylsulfanyl)aniline
Synonyms: O-methioaniline
Molecular Weight (g/mol): 139.22
Molecular Formula: C7H9NS
InChi Key: WBRPQQSADOCKCH-UHFFFAOYSA-N
SMILES: CSC1=CC=CC=C1N
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)