accessibility menu, dialog, popup

UserName

H3CO

CAS RN 2984-50-1

IUPAC Name: 2-hexyloxirane
Synonyms: 2-hexyloxirane
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: NJWSNNWLBMSXQR-UHFFFAOYNA-N
SMILES: CCCCCCC1CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results