accessibility menu, dialog, popup

UserName

KOHOO

CAS RN 298-14-6

IUPAC Name: potassium hydrogen carbonate
Synonyms: potassium bicarbonate
Molecular Weight (g/mol): 100.11
Molecular Formula: CHKO3
InChi Key: TYJJADVDDVDEDZ-UHFFFAOYSA-M
SMILES: [K+].OC([O-])=O
Molecular Weight (g/mol) 
  • (31)
  • (3)
Quantity 
  • (5)
  • (1)
  • (5)
  • (4)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (8)
  • (1)
Grade 
  • (3)
  • (9)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (2)
  • (2)
  • (2)
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (6)
  • (3)
  • (2)
  • (1)
  • (9)
  • (5)
  • (2)
  • (2)
  • (6)
  • (31)
  • (3)
  • (5)
  • (1)
  • (5)
  • (4)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (8)
  • (1)
  • (3)
  • (9)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (5)