accessibility menu, dialog, popup

UserName

CAS RN 29796-57-4

IUPAC Name: triammonium hexachloroiridiumtriuide hydrate
Synonyms: triammonium hexachloroiridiumtriuide hydrate
Molecular Weight (g/mol): 477.05
Molecular Formula: Cl6H14IrN3O
InChi Key: LJHBEBFYNMDGCZ-UHFFFAOYSA-K
SMILES: [NH4+].[NH4+].[NH4+].O.Cl[Ir-3](Cl)(Cl)(Cl)(Cl)Cl
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results